- A direct synthesis of 5,6-dihydroindolo[2,1-a]isoquinolines that exhibit immunosuppressive activity.
A direct synthesis of 5,6-dihydroindolo[2,1-a]isoquinolines that exhibit immunosuppressive activity.
Bioorganic & medicinal chemistry letters (2009-09-05)
George A Kraus, Vinayak Gupta, Marian Kohut, Navrozedeep Singh
PMID19729304
ZUSAMMENFASSUNG
Dihydroindolo[2,1-a]isoquinolines were synthesized from tetrahydroisoquinolines and alpha-fluoroaldehydes by a novel two-step procedure. These compounds exhibited significant immunosuppressive activity against IL-2, IL-10 and IFN-gamma.