Merck
  • Approaches to prolong the use of uncuffed hemodialysis catheters: results of a randomized trial.
Approaches to prolong the use of uncuffed hemodialysis catheters: results of a randomized trial.
American journal of nephrology (2011-03-05)
Vassilis Filiopoulos, Dimitrios Hadjiyannakos, Ioannis Koutis, Sofia Trompouki, Theodora Micha, Dimitrios Lazarou, Dimosthenis Vlassopoulos
ZUSAMMENFASSUNG
Use of uncuffed catheters (UCs) in hemodialysis patients is common practice. An antibiotic lock has been recommended to prevent catheter-related bacteremia (CRB), although insufficient data are available about the appropriate antimicrobial agent and dose with prolonged use of UCs. This open-label randomized study was conducted to compare gentamicin/heparin (group A) and taurolidine/citrate (group B), as catheter-lock solutions, in 119 chronic hemodialysis patients in whom a total of 150 UCs were placed. A well-matched historical control group (heparin) included 67 UCs in 58 patients (group C). CRB episodes developed in 6 and 8 patients in groups A and B, respectively, significantly fewer than in group C (20 patients). Cumulative CRB-free catheter survival at 90 days was 82% for A and 78% for B, which is significantly higher than the 26% for C. Similar Gram-positive infection rates were found in all groups. The Gram-negative infection rate was significantly lower in B compared to C. No significant differences in thrombosis rates were observed between the groups. Gentamicin/heparin and taurolidine/citrate, used for locking UC, were similarly effective at preventing CRB and catheter thrombosis for up to 3 months, until a functional permanent vascular access became available. Both antimicrobial lock solutions were superior to heparin in CRB prevention with similar thrombosis rates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taurolidin, >97% (NMR), powder