Merck
  • cAMP/PKA-dependent increases in Ca Sparks, oscillations and SR Ca stores in retinal arteriolar myocytes after exposure to vasopressin.
cAMP/PKA-dependent increases in Ca Sparks, oscillations and SR Ca stores in retinal arteriolar myocytes after exposure to vasopressin.
Investigative ophthalmology & visual science (2009-12-05)
Owen Jeffries, Mary K McGahon, Peter Bankhead, Maria Manfredi Lozano, C Norman Scholfield, Tim M Curtis, J Graham McGeown
ZUSAMMENFASSUNG
To investigate the effects of arginine vasopressin (AVP) on Ca(2+) sparks and oscillations and on sarcoplasmic reticulum (SR) Ca(2+) content in retinal arteriolar myocytes. Fluo-4-loaded smooth muscle in intact segments of freshly isolated porcine retinal arteriole was imaged by confocal laser microscopy. SR Ca(2+) store content was assessed by recording caffeine-induced Ca(2+) transients with microfluorimetry and fura-2. The frequencies of Ca(2+) sparks and oscillations were increased both during exposure to, and 10 minutes after washout of AVP (10 nM). Caffeine transients were increased in amplitude 10 and 90 minutes after a 3-minute application of AVP. Both AVP-induced Ca(2+) transients and the enhancement of caffeine responses after AVP washout were inhibited by SR 49059, a V(1a) receptor blocker. Forskolin, an activator of adenylyl cyclase, also persistently enhanced caffeine transients. Rp-8-HA-cAMPS, a membrane-permeant PKA inhibitor, prevented enhancement of caffeine transients by both AVP and forskolin. Forskolin, but not AVP, produced a reversible, Rp-8-HA-cAMPS insensitive reduction in basal [Ca(2+)](i). AVP activates a cAMP/PKA-dependent pathway via V(1a) receptors in retinal arteriolar smooth muscle. This effect persistently increases SR Ca(2+) loading, upregulating Ca(2+) sparks and oscillations, and may favor prolonged agonist activity despite receptor desensitization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SR 49059, ≥97% (HPLC)