Merck
Different Effects of Fluoride Exposure on the Three Major Bone Cell Types.
Biological trace element research (2019-03-17)
Ningning Jiang, Fengyang Guo, Boyao Sun, Xiuyun Zhang, Hui Xu
ZUSAMMENFASSUNG
Fluoride accumulates and is toxic to bones. Clinical bone lesions occur in a phased manner, being less severe early in the natural course of skeletal fluorosis. Previous research rarely focused on osteocyte, osteoclast, and osteoblast at the same time, although these three types of cells are involved in the process of fluorosis. In this study, commitment of bone cells was performed according to their respective characteristics. Osteocyte-like cells were verified by protein expression of sclerostin (SOST) in IDG-SW3 cell culture with mineral medium. Positive tartrate-resistant acid phosphatase (TRACP) staining, characteristic of osteoclasts, is observed in RAW264.7 cells after administration of RANKL. We successfully purified a high percentage (94%) of bone mesenchymal stem cells (BMSCs) co-expressing CD34 and CD44. Parallel studies were performed to observe cell viability and apoptosis rates in osteocyte, osteoclast, and osteoblast like cells by using MTT and Annexin V FITC assays. Our results demonstrated that osteocytes have a strong tolerance to high fluoride concentrations, while osteoclasts are more sensitive to changes of fluoride dose. The range of anabolic action of fluoride concentration on osteoblast was narrow. Notably, fluoride exposure aggravated apoptosis of osteocyte and osteoclast induced by administration of PTH and TGF-β, respectively. In short, three types of bone cells display disparate responses to fluoride exposure and to PTH- and TGF-β-induced apoptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
RANK Ligand from mouse, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture