Merck
PDZD8 is a novel Gag-interacting factor that promotes retroviral infection.
Journal of virology (2010-06-25)
Matthew S Henning, Scott G Morham, Stephen P Goff, Mojgan H Naghavi
ZUSAMMENFASSUNG
In a yeast two-hybrid screen for cellular factors that could interact with human immunodeficiency virus type 1 (HIV-1) Gag protein, we identified PDZD8 and confirmed the interaction by coimmunoprecipitation (co-IP). PDZD8 overexpression promoted the initiation of reverse transcription and increased infection by pseudotyped retroviruses independent of the route of viral entry, while transient knockdown of endogenous levels decreased HIV-1 infection. A mutant of PDZD8 lacking a predicted coiled-coil domain in its Gag-interacting region failed to bind Gag and promote HIV-1 infection, identifying the domain of PDZD8 required for mediating these effects. As such, we identify PDZD8 as a novel positive mediator of retroviral infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution