Merck
  • Components of Goutengsan in Rat Plasma by Microdialysis Sampling and Its Protection on Aβ1-42-Induced PC12 Cells Injury.
Components of Goutengsan in Rat Plasma by Microdialysis Sampling and Its Protection on Aβ1-42-Induced PC12 Cells Injury.
Evidence-based complementary and alternative medicine : eCAM (2017-03-30)
Hou-Cai Huang, Chun-Fei Wang, Jun-Fei Gu, Juan Chen, Xue-Feng Hou, Rong-Ling Zhong, Zhi Xia, Di Zhao, Nan Yang, Jing Wang, Xiao-Bin Tan, Xiao-Bin Jia, Liu-Qing Di, Zi-Bo Dong, Liang Feng
ZUSAMMENFASSUNG
Goutengsan, a Chinese herbal formula, potential protection on Alzheimer's disease (AD) has been less reported. In current study, we investigated the protection of Goutengsan on Aβ1-42-induced pheochromocytoma-derived cells (PC12). Furthermore, the components from Goutengsan in rat plasma were identified by microdialysis (MD) for in vivo sampling. Meanwhile, the protection of components identified was also verified. At last, we found that Goutengsan has a potential protective effect on Aβ1-42-induced PC12 cells via reducing cells damage and increasing cells vitality as well as six components (pachymic acid, liquiritin, rhynchophylline, isorhynchophylline, corynoxeine, and isocorynoxeine) which may be effective components. This study helps to understand the treatment of Goutengsan for AD and would facilitate the clinical and further studies for this formula.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rhynchophyllin, ≥95% (HPLC)