Merck
Progard TL1-Vorbehandlungspack Protects the reverse osmosis (RO) membrane from hard water components. For use with Elix® and RiOs™ systems.
Progard® TL1-C Vorbehandlungspack
Progard TL1-Vorbehandlungspack Protects the reverse osmosis (RO) membrane from hard water components. For use with Elix® and RiOs™ systems.
Synonym(e):
Progard® TL1 Vorbehandlungspack