Merck
SU861026
Supelco
Super Clean (Grundplatten-Design) Gasreiniger
triple trap (hydrocarbon, moisture, oxygen), for carrier gas
About This Item
UNSPSC-Code:
41115717
NACRES:
SB.54
Qualitätsniveau
Allgemeine Beschreibung
Super clean (base-plate design) gas purifier is one of the safest ways to help prevent the lowest level of carrier gas contamination from entering the system. The specially designed base-plate has needle valves that instantly close to provide a diffusion-proof seal when the purifier is removed during change out.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Artikel
GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.