Merck
57635
Needle Port Reiniger
About This Item
UNSPSC-Code:
41115854
Allgemeine Beschreibung
Needle port cleaner is an accessory for use with Rheodyne® Model 7725 and 7725i injectors.
Empfohlene Produkte
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Rechtliche Hinweise
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)