Merck
Z762792
Eppendorf® Combitips advanced® pipette tips, Biopur® grade
maximum volume 5.0 mL
Synonym(e):
pipetting tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
blue tip
Sterilität
sterile
Leistungsmerkmale
barrier: no
Verpackung
pack of 100 ea individually wrapped
Hersteller/Markenname
Eppendorf® 0030089669
Maximales Volumen
5.0 mL
Allgemeine Beschreibung
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
The "Biopur" tips are individually blister-wrapped and feature an access tab which makes them easier to open, even with gloves.
Leistungsmerkmale und Vorteile
  • pyrogen-free
  • free of human DNA
  • free of bacterial DNA
  • RNase-free
  • PCR inhibitor-free
  • DNase-free
  • ATP-free
Sonstige Hinweise
Browse all pipette tips
Rechtliche Hinweise
Biopur is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)