Merck
Z742410
PELCO® Sliding Latch Microslide Storage Boxes
yellow, Holds 20 x slides 76x26 mm
Synonym(e):
slide storage
About This Item
UNSPSC-Code:
41123302
NACRES:
MA.01
Materialien
acrylonitrile-butadiene-styrene (ABS)
yellow
Leistungsmerkmale
Holds 20 x slides 76x26 mm
Verpackung
pack of 12 ea
Hersteller/Markenname
Ted Pella 2306-5-CS
L × B × H
104 mm (4.10 in.) × 94 mm (3.70 in.) × 32 mm (1.26 in.)
Allgemeine Beschreibung
Exceptional slide storage containers providing maximum protection for your microscope slides.
Leistungsmerkmale und Vorteile
  • Numbered index sheet in lid for quick identification
  • Foam liner
  • Stackable
  • ABS-durable/impact resistant
  • Hinged lid
  • Unique sliding compression latch
Rechtliche Hinweise
PELCO is a registered trademark of Ted Pella, Inc
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)