Merck
Z741753
3-place collection stand
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
Leistungsmerkmale
capacity three 1.5ml – 2ml tubes
Hersteller/Markenname
IBI ACCFL0003
Allgemeine Beschreibung
The IBI 3-place collection stand is designed to securely hold the flow cell system tubing, luer lock and three 1.5ml – 2ml tubes securely so the biofilm effluent can be collected for analysis.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)