Merck
Z740293
RotoFlex® Plus bench top tube rotator rotisserie
for 1.5 - 2 mL tubes/15 tube capacity per rotisserie
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.73
Verpackung
pack of 2 ea
Hersteller/Markenname
Cole Parmer 439736
Allgemeine Beschreibung
The RotoFlex will accommodate a variety of tubes from 0.5 - 50 mL by simply changing the rotisserie.
Rechtliche Hinweise
RotoFlex is a registered trademark of Argos Technologies, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)