Merck
Z709069
ART® non-filtered tip
HLT 300, size 1-300 μL, racked, non-sterile
Synonym(e):
pipet tips, pipette tips
Größe auswählen
1 PKG
CHF 87.60
CHF 87.60
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 PKG
CHF 87.60
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
CHF 87.60
Check Cart for Availability
Sterilität
non-sterile
Leistungsmerkmale
DNase free
RNase free
racked
Verpackung
pack of 960 ea (10 racks of 96 tips)
Hersteller/Markenname
Molecular BioProducts 3572
Maximales Volumen
300 μL
Größe
1-300 μL
Volumen
1—300 μL
Verwandte Kategorien
Allgemeine Beschreibung
Ideal for Finnpipette and molded with reference marks at 10, 50 and 100 μl.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Rechtliche Hinweise
ART is a registered trademark of Molecular BioProducts, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)