Merck
Z709042
ART® non-filtered tip
HLT 200 Micropoint, size 0.5-200 μL, racked, non-sterile
Synonym(e):
pipet tips, pipette tips
Größe auswählen
1 PKG
CHF 84.30
5 PKG
CHF 298.00
CHF 84.30
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 PKG
CHF 84.30
5 PKG
CHF 298.00
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
CHF 84.30
Check Cart for Availability
Sterilität
non-sterile
Leistungsmerkmale
DNase free
RNase free
racked
Verpackung
pack of 960 ea
Hersteller/Markenname
Molecular BioProducts 3552
Maximales Volumen
200 μL
Größe
0.5-200 μL
Volumen
0.5—200 μL
Verwandte Kategorien
Allgemeine Beschreibung
Micropoint design and 10 μl reference mark for accurate sample handling.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Rechtliche Hinweise
ART is a registered trademark of Molecular BioProducts, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)