Merck
Z659762
Bandelin SONOREX Ultrasonic bath accessories
insert basket for Z659754
Größe auswählen
1 EA
CHF 215.00
CHF 215.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 215.00
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.74
CHF 215.00
Check Cart for Availability
Materialien
stainless steel
Verpackung
pack of 1 ea
Hersteller/Markenname
Bandelin K 10
L × B × T
250 mm × 190 mm × 50 mm
Allgemeine Beschreibung
During ultrasonic operation, nothing may be placed on the tank floor, as this could cause abrasion between the sonication material and the ultrasonic oscillating tank. The use of a suitable basket to hold the sonication material is therefore recommended. This stainless steel baskets is designed so that the sonication material is always in the optimal area of ​​the cavitation field (1-2 cm from the floor) in the oscillating tank. With the help of a basket, the sonication material can also be conveniently placed in the device and removed after operation, e.g. for rinsing, without the user coming into contact with the sonication liquid in the oscillating tank.
Leistungsmerkmale und Vorteile
  • perfect positioning in the sound field
  • optimal loading and unloading
Rechtliche Hinweise
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)