Z374563
Eppendorf® Multipette® Plus pipette
Synonym(e):
Adjustable volume pipette
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
Empfohlene Produkte
Leistungsmerkmale
electronic: no
single-channel
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 022260201
Eppendorf® 4981 000.019
Allgemeine Beschreibung
Accepts any of seven interchangeable combitips; each tip gives a choice of 20 preset volumes. The combitip′s integral reservoir lets you pipette up to 100 times without refilling. Pipette automatically recognizes tip and digitally displays selected volume.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)