Merck
TMO331840-BR
Nunc® barcoded latch rack
for low profile 300 mL external thread universal tubes, plastic, 30 positions, case of 10 ea
Synonym(e):
tube rack, tube storage
Größe auswählen
10 EACH
CHF 313.00
CHF 313.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EACH
CHF 313.00
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.12
CHF 313.00
Check Cart for Availability
Materialien
plastic
Verpackung
case of 10 ea
Hersteller/Markenname
Nunc 331840-BR
Allgemeine Beschreibung
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific Nunc Universal Latch Rack for Cryogenic Tubes.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)