Merck
TMO312004-9125
Nalgene® diagnostic bottle
amber high-density polyethylene, volume 4 mL, case of 2000 ea
Synonym(e):
PPCO bottles, small volume bottle with closure
Größe auswählen
2000 EACH
CHF 1’550.00
CHF 1’550.00
Warenkorb auf Verfügbarkeit prüfen
Größe auswählen
Ansicht ändern
2000 EACH
CHF 1’550.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 1’550.00
Warenkorb auf Verfügbarkeit prüfen
Materialien
amber high-density polyethylene
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 312004-9125
Kapazität
4 mL
Volumen
4 mL
Allgemeine Beschreibung
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)