T5026
Drummond sequencing pipette tips
O.D. 0.19 mm
Alle Fotos(1)
About This Item
Materialien
polycarbonate
Sterilität
non-sterile; autoclaved
Verpackung
pack of 10 ea
Hersteller/Markenname
Drummond Scientific 3-000-203-G/.2
AD
0.19 mm
Allgemeine Beschreibung
Disposable, autoclavable, flexible, polycarbonate pipet tips.