Merck
PURDR
Sigma-Aldrich
Pur-A-Lyzer Floating Racks for Dialysis
holds up to 4 Midi Pur-A-Lyzer dialysis tubes, pack of 1 ea
Synonym(e):
Dialyzers, High through put dialysis tubing
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.24
Verpackung
pack of 1 ea
Allgemeine Beschreibung
Floating rack can hold up to 4 Midi Pur-A-Lyzer Dialysis Tubes in a single beaker.
Rechtliche Hinweise
Pur-A-Lyzer is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)