OH30240974
Ohaus® pH Electrode
ST270
Alle Fotos(1)
About This Item
UNSPSC-Code:
41115604
Empfohlene Produkte
Materialien
epoxy shaft
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30240974
pH-Messbereich
0.00-14 pH unit
Temperaturmessbereich
0.0-100 °C
Allgemeine Beschreibung
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Anwendung
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Leistungsmerkmale und Vorteile
Can be used in conjunction with all Starter bench and portable meters
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)