Merck
MTCP520020U
MTC Bio ProPette LE Adjustable Pipettor
capacity 2-20 μL
Synonym(e):
pipette
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.15
Leistungsmerkmale
adjustable
manual
no. of channels 1
Verpackung
pack of 1 ea
Hersteller/Markenname
MTC Bio P5200-20U
Kapazität
2-20 μL
Verwandte Kategorien
Allgemeine Beschreibung
ProPette LE Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette LE, 2 to 20µL
Leistungsmerkmale und Vorteile
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (0.1µL-10mL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Rechtliche Hinweise
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)