Merck
M9064
Sigma-Aldrich
Thumbs-Up microtubes
volume 1.5 mL
Synonym(e):
plastic vials
About This Item
UNSPSC-Code:
41121702
NACRES:
NB.22
Volumen
1.5 mL
Allgemeine Beschreibung
Saves fingernails from breaking and cracking. Opens with a flip of a thumb. Reduces sample contamination. Fast and easy removal from Rotors. Inner lid structure allows for pre-loading of reactants. RNase and DNase free. Fully autoclavable propylene. RCF rating of 17,000 × g. Can withstand temperatures ranging from - 196°C to 130°C.
Rechtliche Hinweise
Thumbs-Up is a trademark of Diversified Biotech
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)