Merck
GN791840
Greiner 3-D Aldehyde HTA plate
non-sterile, 96 tests per plate
About This Item
UNSPSC-Code:
41121800
Verpackung
pack of 5 ea
Hersteller/Markenname
Greiner 791840
Verfügbarkeit
available only in China
Größe
96 wells
Eignung
suitable for (cell culture)
Allgemeine Beschreibung
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA Plate has been optimized for the simultaneous analysis of multiple samples.
Leistungsmerkmale und Vorteile
  • low well rim of only 0.3 mm
  • low autofluorescence
Rechtliche Hinweise
HTA is a trademark of Greiner Bio-One GmbH
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)