Merck
GN701061
Greiner Multiwell Strips
high binding, wells flat bottom (1 × 12 well)
Synonym(e):
Greiner Multiwell Strips
About This Item
UNSPSC-Code:
41121800
Verpackung
pack of 400 ea
Hersteller/Markenname
Greiner 701061
Verfügbarkeit
available only in China
Größe
12 well
Bindungsart
high binding surface
Allgemeine Beschreibung
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)