Merck
GN381070
Greiner CapMat for sealing multiwell plates
96 well, round nap, ethylene vinyl acetate, translucent
Synonym(e):
Greiner CapMats for sealing multiwell plates
About This Item
UNSPSC-Code:
41122410
Verpackung
pack of 50 ea
Hersteller/Markenname
Greiner 381070
Verfügbarkeit
available only in China
Chemische/physikalische Beständigkeit
Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C