Merck
GE80-6466-65
GE Ettan DALT precast gel cassette
gel size 26 cm × 20 cm × 0.1 cm
About This Item
UNSPSC-Code:
41105300
NACRES:
NB.22
Verpackung
pack of 1 ea
Hersteller/Markenname
Cytiva 80-6466-65
Cytiva Article No. 80-6466-65
Gelgröße
26 cm × 20 cm × 0.1 cm
Allgemeine Beschreibung
Simple-to-use gel cassettes for gel loading, unloading, and storage. To be used with DALT is an integrated system for reproducible high resolution, high throughput 2-D electrophoresis.
Rechtliche Hinweise
Ettan is a trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
This protocol shows how to insert gels into Ettan™ DALT electrophoresis units.
Verwandter Inhalt
This page shows how to apply equilibrated Immobiline DryStrip gels