Merck
GBL666283
Grace Bio-Labs Press-To-Seal silicone isolator, red silicone sheet
thickness 1.8 mm
Synonym(e):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
Größe auswählen
5 EA
CHF 488.00
CHF 488.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
5 EA
CHF 488.00
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.22
CHF 488.00
Check Cart for Availability
Materialien
red silicon
Leistungsmerkmale
(Adhesive on both sides)
Verpackung
pack of 5 ea
Hersteller/Markenname
Grace Bio-Labs 666283
L × B
180 mm × 130 mm
Dicke
1.8 mm
Allgemeine Beschreibung
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)