Merck
EP6731010215
Eppendorf® CellXpert® C170i CO2 Incubator
new
water monitoring, inner door non-segmented, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
Synonym(e):
Heaters
About This Item
UNSPSC-Code:
41104405
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 6731010215
Allgemeine Beschreibung
CellXpert® C170i, non-segmented inner door, handle right side, water-level monitoring, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Leistungsmerkmale und Vorteile
  • water-level monitoring
  • handle right side
  • non-segmented inner door
  • 180° HTD (standard with C170i)
  • including water tray and four shelves (stainless steel)
  • 100 – 127 V, 50 – 60 Hz
Rechtliche Hinweise
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)