Merck
EP5920761005
Eppendorf® Adapter for 4 x 15ml conical tubes
for FA-6x250 rotor, case of 2 ea
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.78
Leistungsmerkmale
for FA-6x250 rotor
Verpackung
case of 2 ea
Hersteller/Markenname
Eppendorf® 5920761005
Parameter
10,000 rpm speed
Lagertemp.
room temp
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG