Merck
EP5409704001
Rotor for Eppendorf® 5427R Centrifuge
FA-45-24-11-Kit, aerosol tight, Eppendorf® 5409704001 (Global), lid
Größe auswählen
1 EA
CHF 998.00
CHF 998.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 998.00
About This Item
UNSPSC-Code:
41103909
NACRES:
NB.82
CHF 998.00
Check Cart for Availability
Leistungsmerkmale
lid
to hold 24 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 5409704001 (Global)
Parameter
13,200 rpm
19,100 × g max. RCF
Methode(n)
centrifugation: suitable
Kompatibilität
for use with centrifuge models 5427R
Allgemeine Beschreibung
Rotor incudesan aerosol-tight lid and is to be used with Eppendorf centrifuge 5427 R. It has a maximum spinning speed of 19090 × g (13,200 rpm). Aerosol tightness of lid has been tested and certified in accordance with Annex AA of the IEC 1010-2-020/IEC 61010-2-020. Certificate is included in the operating manual.
Rotors to fit Eppendorf 5427 R refrigerated centrifuge.
Leistungsmerkmale und Vorteile
  • Max. speed: 19,090 × g (13,200 rpm)
  • Rotor with higher rim for safe centrifugation of up to 24 spin columns
  • Aerosol-tight with Eppendorf QuickLock lid
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)