Merck
EP4920609002
Eppendorf® Reference 2 lower part
single-channel, lower part with separate serial number, red, size 0.25-2.5 mL
Größe auswählen
1 EA
CHF 67.50
CHF 67.50
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 67.50
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
CHF 67.50
Check Cart for Availability
Leistungsmerkmale
single-channel
lower part with separate serial number
Verpackung
pkg of 1 ea
Hersteller/Markenname
Eppendorf® 4920609002
Größe
0.25-2.5 mL
Farbe
red
Allgemeine Beschreibung
The name, Reference, stands for extraordinary precision and accuracy, a long service lifes and an ergonomic design. The new Reference 2 boasts these proven premium characteristics with its state-of-the-art technolgy, making it a reliable partner for you and your work. The time saving single-button operation is unrivalled in the new multichannel versions.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)