Merck
CLS4320C100
Pyrex® round-bottom heavy wall boiling flask, short neck with joint
capacity 100 mL, joint: ST/NS 24/40
Synonym(e):
Corning® 4320C
About This Item
UNSPSC-Code:
41121804
Materialien
round bottom flask
Verbindung
joint: ST/NS24/40
Verpackung
case of 1 ea
Hersteller/Markenname
Corning 4320C-100
Kapazität
100 mL
Halsumfang
(1)
Allgemeine Beschreibung
Pyrex heavy duty boiling flask provides increased mechanical strength. The flask features walls that are 25-30% thicker than the standard version.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)