Merck
CLS3772
Corning® microcarriers
non-treated
, weight 10 g
Synonym(e):
cell culture
Größe auswählen
1 EA
CHF 95.30
CHF 95.30
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 95.30
About This Item
UNSPSC-Code:
41122100
NACRES:
NB.13
CHF 95.30
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Corning 3772
Gewicht
10 g
Allgemeine Beschreibung
Corning Microcarriers are designed to simplify your scale-up processes. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors.
Features:
  • USP Class VI polystyrene material provides a consistent platform
  • Level (SAL) of 10-6
  • Certified nonpyrogenic
These microcarriers are untreated for attachment of extra sticky cells or for alternate coatings.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1