Merck
CLS3584
Corning® volume adapter for 96 well filter plates
metal, non-sterile
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.24
Materialien
metal
Sterilität
non-sterile
Verpackung
pack of 10 ea
case of 50 ea
Hersteller/Markenname
Corning 3584
Well-Volumen
1 mL
Wells
96 wells
Allgemeine Beschreibung
Volume adapter mounts on top of the 96 well FiltrEX; filter plate and allows larger volumes (1mL) to be applied to each well in a filter plate. Adapters are nonsterile.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)