BR31577
BRAND® BLAUBRAND® measuring cylinder with serialized numbers
capacity 1000 mL
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
Verpackung
pkg of 12 ea
Hersteller/Markenname
BRAND 27452
Höhe
115 mm
Kapazität
1000 mL
Allgemeine Beschreibung
Measuring cylinders tall form. Class A DE-M marking to DIN EN ISO 4788. Made from DURAN(R), calibrated to contain (TC, In). With spout and hexagonal glass base. Ring marks at major graduations. High contrast blue enamel graduations and inscription
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)