Merck
BMSW10052
Sigma-Aldrich
Accuris M1 Grade Calibration Weight
size 2 g, stainless steel
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.71
Materialien
stainless steel
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific W1005-2
Größe
2 g
Allgemeine Beschreibung
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)