Wichtige Dokumente
NU050/441-322
Millipore
NovAseptic® Tank outlet (1/2") Valve Body 90°
Valve Body - ASTM® A269/270 and Outlet with Elbow 90 °, Electro-polished with TC connection
About This Item
UNSPSC-Code:
23151800
Qualitätsniveau
Parameter
-1-10 bar max. pressure (-14.5-145 psig) at 25 °C (atmospheric pressure)
-80-200 °C max. temp. (-112-392 °F)
Allgemeine Beschreibung
NovAseptic® Valves are engineered for optimal performance, reliability, durability and ease of maintenance. NovAseptic® Valves comply with cGMP Design Qualification
criteria for aseptic processing. This is why we choose to call it ‘‘Aseptic by Design.”
Spezifität
Surface roughness: Internal surface Ra ≤ 0.38 μm (15 μin) External surface Ra ≤ 1.6 μm (63 μin)
Anwendung
NovAseptic® Valves are specifically designed for aseptic applications and comply with the most stringent cleanability and sterilizability requirements. The focus on aseptic design is a significant feature for all valves. Flow through valve is a valve designed for a wide range of applications in the aseptic process; including manifolds, take off and drainage valves.
Komponenten
All open ends on the valve body are protected with plastic covers on delivery and packaged in a closed box.
Rechtliche Hinweise
ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany