Merck
Z743108
Julabo® flow through immersion cooler
FD 200, temp. range 10 °C (30 °C), AC/DC input 115 V AC, US 3-pin plug (5-15P)
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Verpackung
pack of 1 ea
Hersteller/Markenname
FD 200
Julabo 9655825
Parameter
10 °C temp. range (30 °C)
Allgemeine Beschreibung
JULABO flow-through coolers are the perfect complement to open bath circulators, immersion circulators and bridge mounted circulators extending their temperature range to below ambient temperature. They offer a small footpri nt, have a low energy requirement and represent an economic solution preventing tap water usage for cooling. Instruments are designed for integration into an existing cooling loop (e.g. consisting of open bath circulator wit h pump).
Your advantages
  • For applications near ambient temperatures
  • Integrated freezing protection
  • Preventing the use of precious tap water for cooling
  • Compact design, small footprint
  • Ease of operation
  • Low energy requirement
  • Time saving - Comfortable and simple operation for setpoint adjustment
  • Precise Temperature Control - PID Temperature control with set control parameters, temperature stability ±0.02...±0.2 °C
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)