Merck
Z742598
Accuris SmartDoc narrow band pass filter
Orange PMMA; Orange photo filter; Used for imaging green stains
Synonym(e):
Accuris SmartDoc
Größe auswählen
1 EA
CHF 35.50
CHF 35.50
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 35.50
About This Item
UNSPSC-Code:
41106000
NACRES:
NB.79
CHF 35.50
Check Cart for Availability
Leistungsmerkmale
Orange PMMA; Orange photo filter; Used for imaging green stains
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific E5001-ORANGE
Allgemeine Beschreibung
An orange photo filter used to image green stains on blue light illuminators and transilluminators.
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)