Merck
Z742262
DURAN® TILT Light Shield
white silicone, includes GL 56 bottle tags
Synonym(e):
292435601
Größe auswählen
4 EA
CHF 34.90
CHF 34.90
Check Cart for Availability
Größe auswählen
Ansicht ändern
4 EA
CHF 34.90
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 34.90
Check Cart for Availability
Materialien
white silicone
Leistungsmerkmale
includes GL 56 bottle tags
Verpackung
pack of 4
Hersteller/Markenname
DURAN® 292435601
Allgemeine Beschreibung
The DURAN TILT light shield is a white silicone sleeve that covers 94% of the bottle surface. The sleeve has a number of protective functions. It blocks damaging UV light, protects the surface from damage, and facilitates safer handling. Silicone provides a much better grip than glass, especially when the bottle has been sprayed with 70% alcohol for surface disinfection.
Leistungsmerkmale und Vorteile
  • Manufactured from silicone
  • The white DURAN® TILT Light Shield is a silicone sleeve that covers 94 % of the bottle surface and blocks damaging ultraviolet light
  • Protects the glass surface from damage
  • Facilitates safer handling
  • Silicone provides a much better grip than glass especially when the bottle has been sprayed with 70 % alcohol for surface disinfection
Sonstige Hinweise
Includes four GL 56 Bottle Tags - orange, yellow, blue, and purple
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)