Merck
Z690686
BactiZapper slide dryer attachment for 3 slides
aluminum holder (with a stainless steel clip)
Größe auswählen
1 EA
CHF 157.00
CHF 157.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 157.00
About This Item
UNSPSC-Code:
41103200
NACRES:
NB.13
CHF 157.00
Check Cart for Availability
Materialien
aluminum holder (with a stainless steel clip)
Hersteller/Markenname
BactiZapper
Benchmark Scientific B1000-03
Allgemeine Beschreibung
The BactiZapper slide dryer is an aluminum holder that attaches to the BactiZapper infrared sterilizer (Z690651, 230 V or Z690643, 115 V) to dry laboratory slides. The unit connects to the sterilizer with a stainless steel clip and uses heat radiating out of the device to dry laboratory slides.
The slide dryer holds three slides for drying multiple samples per use.
Rechtliche Hinweise
BactiZapper is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)