Merck
Z684570
Ace Rotary Evaporator Vial Adapter Kit
Chemraz O-ring, adapter joint: ST/NS 29/32
Größe auswählen
1 EA
CHF 869.00
CHF 869.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 869.00
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
CHF 869.00
Check Cart for Availability
Materialien
Chemraz O-ring
Verbindung
joint: ST/NS adapter29/32
Beschreibung
PTFE adapters with #15 Ace-Thred to GPI vial thread, sizes 8-425, 13-425, 15-425, 18-400, 20-400, 22-400, 24-410
Hersteller/Markenname
Ace Glass 13290-69
Verwandte Kategorien
Allgemeine Beschreibung
PTFE connecting adapters and glass conversion adapters for connecting GPI threaded vials to a rotary evaporator vapor tube or trap. Both are suitable for vacuum work. PTFE connecting adapters are available with FETFE® or Chemraz® O-rings. Order Chemraz if you are using methylene chloride or acetone.
Rechtliche Hinweise
Ace-Thred is a trademark of Ace Glass, Inc.
Chemraz is a registered trademark of Greene, Tweed of Delaware, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)