Merck
Z674362
Duran® GLS 80 caps and accessories
GL 18 blue screw cap for tube connection
Synonym(e):
1160174
Größe auswählen
2 EA
CHF 16.60
CHF 16.60
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
CHF 16.60
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 16.60
Check Cart for Availability
Hersteller/Markenname
DURAN® 1160174
Allgemeine Beschreibung
Flexible modular liquid or gas transfer 80 connection cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
Leistungsmerkmale und Vorteile
  • Manufactured from PP, PTFE, EPDM
  • The flexible modular system allows an individual combination of the different components according to requirements and application
  • Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts
  • Sterile pressure equalisation is possible by using the syringe filter
  • Unused ports can be sealed off with a blank GL 18 screw cap
  • Fully compatible with all the GLS 80 threads and therefore highly versatile
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)