Merck
Z673757
Hanna HI 3000 series bench pH/mV/Temp/ISE meters
1 input channel, HI 3220-02, AC/DC input 230 V AC, EuroPlug
About This Item
UNSPSC-Code:
41115603
Hersteller/Markenname
Hanna HI 3220-02
Messbereich mV
±2000 mV, accuracy: 0.2 °C
pH-Messbereich
-2.0-20.0, accuracy: ±0.1
-2.00-20.00, accuracy: ±0.01
-2.000-20.000, accuracy: ±0.2
Temperaturmessbereich
-20-120 °C, accuracy: ±0.2 °C
Allgemeine Beschreibung
Hanna HI 3000 series bench meters measure pH, mV, temp and ISE. Equipped with the exclusive Calibration Check Electrode Diagnostics to alert the user when the electrode needs cleaning. Prompts on the graffic LCD makes callibration easy and accurate.
Supplied with HI 1131B electrode with BCN connector and 1 m cable, HI 7662-T SS temperature probe, pH 4 and 7 buffer solutions, cleaning solution, electrolyte solution, electrode holder, 12 V DC power adapter and instructions.
For UK version please order European 2-pin plug product plus Z673978 (UK 3-pin power cord).
  • Calibration Check Electrode Diagnostics
  • Callibration prompts on graffic LCD
  • 5 point calibration with 7 standard and 5 custom buffers
  • Relative mV measurements
  • Log on demand for up to 400 samples
  • Auto HOLD to freeze readings on the display
  • GLP to view last calibration data for pH, Rel mV or ISE
  • User selectable "Outside Calibration Range" warning
  • User-selectable "calibration time out" to remind when new calibration is necessary
  • PC compatible via USB
Sonstige Hinweise
Without ISE
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)