Merck
Z673501
LAB-AIRE® II Trockengestelle, nicht elektrisch
benchtop, single sided, 2 tier
Größe auswählen
1 EA
CHF 299.00
CHF 299.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 299.00
About This Item
UNSPSC-Code:
41103200
NACRES:
NB.35
CHF 299.00
Check Cart for Availability
Leistungsmerkmale
no. of pegs 38
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F18933-0012
L × B × H
14.75 in. × 7 in. × 15.4 in.
Allgemeine Beschreibung
Space in laboratories is always in high demand making the small footprint of the Scienceware® Lab-Aire II Drying Racks a practical solution for drying and storing labware. With a choice of benchtop or wall-mountable models in varying heights, you′re sure to find the rack that meets your specific needs.
All models come with three size and length rugged V-channel pegs that can be moved and repositioned to hold a variety of labware, including: beakers, flasks, bottles, test tubes and cylinders.
Additional accessory pegs, (19 per bag, small, medium or large) are available to reconfigure the rack for specific size and weight labware. Accommodates an assortment of glass and plastic labware by easily changing peg locations.
  • High storage capacity - up to 114 peg positions for all types of labware
  • Single-sided benchtop models fit against a wall, conserving bench space
  • Double-sided benchtop models provide twice the capacity with only a slightly larger footprint
  • Wall-mount models to save bench space
Rechtliche Hinweise
Lab-Aire is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)