Merck
Z557609
Sigma-Aldrich
Aldrich® pear-shaped splash-guard adapter
capacity 250 mL, female joint: ST/NS 24/40, male joint: ST/NS 24/40
Größe auswählen
1 EA
CHF 181.00
CHF 181.00
Voraussichtliches Versanddatum26. April 2025
Größe auswählen
Ansicht ändern
1 EA
CHF 181.00
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
CHF 181.00
Voraussichtliches Versanddatum26. April 2025
Verbindung
female joint: ST/NS24/40
male joint: ST/NS24/40
Beschreibung
anti-climb
Kapazität
250 mL
Allgemeine Beschreibung
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)