Merck
Z282421
Sigma-Aldrich
Spinbar® Sortiment
polygon PTFE-coated, pack of 18 ea
About This Item
UNSPSC-Code:
41103806
NACRES:
NB.72
Materialien
polygon PTFE-coated
Verpackung
pack of 18 ea
Hersteller/Markenname
Bel-Art F37184-0000
Allgemeine Beschreibung
2 of each size: 10 × 6 mm, 15 × 6 mm, 20 × 7 mm, 25 × 8 mm, 30 × 8 mm, 40 × 8 mm, 50 × 8 mm, 60 × 8 mm, 80 × 10 mm
Rechtliche Hinweise
Spinbar is a registered trademark of SP Scienceware, a division of SP