Merck
Z269603
Safety pipette fillers
green
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
natural rubber
Hersteller/Markenname
FLIP-004
Farbe
green
Allgemeine Beschreibung
Has two operating valves to fill and dispense and a third removable automatic valve at the top for evacuation and easy inside cleaning. Natural rubber.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)